Learn More
Mettler Toledo™ InLab™ Sensor Cables: S7 Connection
Product Code. 15569661 Shop All Mettler Toledo Products
Click to view available options
Connector Types:
S7-BNC
S7-BNC, IP67 waterproof
S7-DIN 19262
S7-Lemo 00
S7-None
Length (Metric) Cable:
1.2 m
10 m
3.0 m
5.0 m
Unit Size:
Each
Product Code. 15569661
Brand: Mettler Toledo™ 30281925
Electrode cables to connect Mettler Toledo InLab sensors to any Mettler Toledo meter.
  • Detachable
  • Resuable
TRUSTED_SUSTAINABILITY
Specifications
Connector Types S7-Lemo 00
For Use With (Equipment) Mettler Toledo InLab Sensors
For Use With (Application) To connect Mettler Toledo InLab sensors to any meter
Length (English) Cable 4 ft.
Length (Metric) Cable 1.2 m