Learn More
Mettler Toledo™ InLab™ Sensor Cables: For Temperature Probes
Product Code. 15559661 Shop All Mettler Toledo Products
Change view
Click to view available options
Connector Types:
RCA (Cinch)
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connector Types unitSize
15559661 RCA (Cinch) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15559661 Supplier Mettler Toledo™ Supplier No. 30281924
Add to basket
Add to basket
Electrode cables to connect Mettler Toledo Temperature Probes to any meter.
  • Detachable
  • Resuable
TRUSTED_SUSTAINABILITY
Specifications
Connector Types RCA (Cinch)
For Use With (Equipment) Mettler Toledo Temperature Probes
For Use With (Application) To connect Mettler Toledo temperature probes to any meter
Length (English) Cable 4 ft.
Length (Metric) Cable 1.2 m