Learn More
Mettler Toledo™ InLab™ 605 ISM Intelligent Dissolved Oxygen Sensors
Brand:  Mettler Toledo™ 51344613
1030.75 EUR valid until 2023-12-31
Use promo code "18562" to get your promotional price.
Additional Details : Weight : 0.47000kg
 View more versions of this product
Product Code. 10126064
  • 1085.00€ / Each
Estimated Shipment: 24-11-2023
Specifications
10 m
0°C to 60°C
CE, ISO 9001 certified