Learn More
B Braun™ Injekt Solo Cone Syringes
Click to view available options
Volume (Metric):
2 mL
10 mL
20 mL
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 100
Pack of 1000
Description
Enjoy the smooth, steady operation of the B Braun™ Solo Cone Leur syringe. Its green plunger enables easy recognition of filling volumes and its double-seal stopper keeps contents intact.
- Injekt™
- Transparent barrel with graduations
- Green plunger
- Barrel and plunger rod: Polypropylene
- No DEHP or natural rubber
Specifications
Specifications
| Certifications/Compliance | EN ISO 7886-1 |
| Product Type | Syringe |
| Disposable | Single-use |
| Quantity | 1/Ea. |
| Volume (Metric) | 20 mL |
| Sterility | Sterile |
Product Content Correction
Product Title
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering