Learn More
Ingenol, TRC
Specifications
Specifications
| Chemical Name or Material | Ingenol |
| CAS | 30220-46-3 |
| Molecular Formula | C20H28O5 |
| Quantity | 50 mg |
| Synonym | (1aR,2S,5R,5aR,6S,8aS,9R,10aR)-1a,2,5,5a,6,9,10,10a-Octahydro-5,5a,6-trihydroxy-4-(hydroxymethyl)-1,1,7,9-tetramethyl-1H-2,8a-methanocyclopenta[a]cyclopropa[e]cyclodecen-11-one, [1aR-(1aα,2β,5β,5aβ,6β,8aα,9α,10aα)]-1a,2,5,5a,6,9,10,10a-Octahydro-5,5a,6-trihydroxy-4-(hydroxymethyl)-1,1,7,9-tetramethyl-1H-2,8a-methanocyclopenta[a]cyclopropa[e]cyclodecen-11-one |
| InChI Formula | InChI=1S/C20H28O5/c1-9-7-19-10(2)5-13-14(18(13,3)4)12(17(19)24)6-11(8-21)16(23)20(19,25)15(9)22/h6-7,10,12-16,21-23,25H,5,8H2,1-4H3/t10-,12+,13-,14+,15+,16-,19+,20-/m1/s1 |
| SMILES | O[C@H]1C(C)=C[C@@]2([C@]1(O)[C@H](O)C(CO)=C[C@@]3([H])[C@@](C4(C)C)([H])[C@@]4([H])C[C@H]2C)C3=O |
| Molecular Weight (g/mol) | 348.43 |
| Formula Weight | 348.1937 |