missing translation for 'onlineSavingsMsg'
Learn More

ING5 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Thermo Scientific PA560081

Additional Details : Weight : 0.02000kg

Product Code. 15747921

  • 514.00€ / 100µL
Estimated Shipment: 10-05-2024
to see stock.
Add to basket

Description

Description

Immunogen sequence: EDKKAEIDIL AAEYISTVKT LSPDQRVERL QKIQNAYSKC KEYSDDKVQ Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 92%.

ING5 is similar to ING1, a tumor suppressor protein that can interact with TP53, inhibit cell growth, and induce apoptosis. This protein contains a PHD-finger, which is a common motif in proteins involved in chromatin remodeling. This protein can bind TP53 and EP300/p300, a component of the histone acetyl transferase complex, suggesting its involvement in TP53-dependent regulatory pathway.
Specifications

Specifications

ING5
Polyclonal
Unconjugated
ING5
1700001C14Rik; 1700027H23Rik; 1810018M11Rik; AI225768; ING5; inhibitor of growth family member 5; inhibitor of growth family, member 5; inhibitor of growth protein 5; p28ING5
Rabbit
Antigen affinity chromatography
RUO
84289
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q8WYH8
ING5
Recombinant protein corresponding to Human ING5
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Research Use Only