Learn More
Fisherbrand™ Infrared Thermometer
Product Code. 12113811 Shop All Fisherbrand Products
Change view
Click to view available options
Temperature Range (Metric):
-33°C to +500°C
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature Range (Metric) unitSize
12113811 -33°C to +500°C Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12113811
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Specifications
Depth (Metric) 39 mm
Dimensions (L x W x H) 178 H x 79 L x 39 D mm
Emissivity 0.95
Height (Metric) 178 mm
Humidity Range 80RH
Length (Metric) 79 mm
Length (Metric) Overall 79 mm
Resolution 0.1°C
Weight (Metric) 170 g
Product Type Infrared-Thermometer
Max. Temperature (Metric) 500°C
Min. Temperature (Metric) -33°C
Temperature Range (Metric) -33°C to +500°C