missing translation for 'onlineSavingsMsg'
Learn More

Indoxyl Beta-D-Glucuronide CHA Salt, IBDG, TRC

Product Code. 30390818
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30390818 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30390818 Supplier Toronto Research Chemicals Supplier No. TRCI65410850MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 216971-58-3
Molecular Formula C14H15NO7 . C6H13N
Quantity 50 mg
Synonym 1H-Indol-3-yl β-D-Glucopyranosiduronic Acid compd. with Cyclohexanamine
InChI Formula InChI=1S/C14H15NO7.C6H13N/c16-9-10(17)12(13(19)20)22-14(11(9)18)21-8-5-15-7-4-2-1-3-6(7)8;7-6-4-2-1-3-5-6/h1-5,9-12,14-18H,(H,19,20);6H,1-5,7H2/t9-,10-,11+,12-,14+;/m0./s1
SMILES NC1CCCCC1.O[C@H]([C@@H](O)[C@@H]2O)[C@@H](O[C@@H]2C(O)=O)OC3=CNC4=C3C=CC=C4
IUPAC Name cyclohexanamine (2S,3S,4S,5R,6S)-6-((1H-indol-3-yl)oxy)-3,4,5-trihydroxytetrahydro-2H-pyran-2-carboxylate
Molecular Weight (g/mol) 408.44
Formula Weight 408.18
Chemical Name or Material Indoxyl beta-D-Glucuronide CHA Salt, IBDG
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.