Learn More
Indole-4-boronic acid, 95%
Product Code. 15469807
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. 15469807
Brand: Thermo Scientific Alfa Aesar H53037.MD
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
CAS: 220465-43-0 | C8H8BNO2 | 160.967 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 220465-43-0
Molecular Formula C8H8BNO2
Molecular Weight (g/mol) 160.967
MDL Number MFCD03095175
InChI Key USJUQEVUEBCLLR-UHFFFAOYSA-N
Synonym indole-4-boronic acid, 4-boronoindole, 1h-indol-4-yl boronic acid, 1h-indole-4-boronic acid, 4-indoleboronic acid, 4-indolylboronic acid, 4-borono-1h-indole, 1h-indol-4-yl-4-boronic acid, boronic acid, 1h-indol-4-yl, b-1h-indol-4-yl-boronic acid
PubChem CID 2763173
IUPAC Name 1H-indol-4-ylboronic acid
SMILES B(C1=C2C=CNC2=CC=C1)(O)O
Specifications
Quantity 250 mg
Sensitivity Air Sensitive
Formula Weight 160.97
Percent Purity 95%
Chemical Name or Material Indole-4-boronic acid