Learn More
Fisherbrand™ Individual Stainless Steel Cylindrical Calibration Weights, Class E2 with Certificate
Product Code. 15575069 Shop All Fisherbrand Products
Click to view available options
Weight (Metric):
1 g
5 g
10 g
20 g
50 g
100 g
200 g
500 g
1 kg
2 kg
5 kg
10 kg
Unit Size:
1 unit
Product Code. 15575069
Calibrate weights of lower accuracy classes. Fisherbrand™ Class E2 Individual Stainless Steel Cylindrical Calibration Weights are intended for use in the verification or calibration of class F1 weights and for use with weighing instruments of special accuracy class I.
  • Class E2 stainless steel calibration weight
  • In compliance with OIML R111
  • Delivered with a calibration certificate guaranteeing traceability
Specifications
Classification E2
Material Stainless Steel
Weight (Metric) 1 kg
Mass 1 kg
Shape Cylindrical