Learn More
Inconel™ 600 gauze, 60 mesh woven from 0.19mm (0.0075 in.) dia. wire, Thermo Scientific™
Product Code. p-7080148
Click to view available options
Quantity:
75 x 75 mm
150 x 150 mm
300 x 300 mm
Unit Size:
Each
Product Code. 15497255
Brand: Thermo Scientific Alfa Aesar 046584.FL
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Inconel™ 600 gauze
Name Note 60 mesh woven from 0.19mm (0.0075 in.) dia. wire
Avg. Mol. Wt. or Mol. Wt. Range Ni:Cr:Fe; 77:15.5:7.5 wt%
Quantity 75 x 75 mm
Mesh Size 60 Mesh
Form Wire Cloth
MDL Number MFCD01741927