missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX InCELL Hunter™ HEK 293 RIPK2 Kinase Binding Cell Line
Description
The InCELL Hunter™ HEK 293 RIPK2 Kinase Binding Cell Line is a stable, engineered cell line for use in studying small molecule drug candidates that engage with kinases. This cell line enables assessment of compound-engagement-based stabilization of RIPK2 via detection of RIPK2 levels. This product uses EFC technology. The cell line included overexpresses RIPK2 target protein fused to PL. In the absence of compound that binds and stabilizes the target, this protein is denatured by the heat treatment and no EFC signal is produced. When compound engages and heat-stabilizes the target, EA contained in the detection reagent produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells.
Specifications
Specifications
| Assay Entry | Target Engagement |
| Cell Line | 293H |
| For Use With (Application) | Cell Lines |
| Primary Agonist | Foretinib (GSK1363089) |
| Quantity | 2 Vials |
| Target Entry | RIPK2 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?