missing translation for 'onlineSavingsMsg'
Learn More

DiscoverX InCELL Hunter™ eXpress Bcl2L1-Bax Protein Binding Assay

Product Code. 17314769
Encompass
Click to view available options
Quantity:
1,000 dp (10 x 96-well)
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 17314769

Brand: DiscoverX 960009E3CP16L

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The InCELL Hunter™ eXpress Bcl2L1-Bax Protein Binding Assay measures protein binding via compound-induced binding partner disruption and degradation. eXpress kits contain all assay materials: cells, reagents, and plates.

The InCELL Hunter™ eXpress Bcl2L1-Bax Protein Binding Assay provides an easy-to-use cell based assay to study small molecule drug candidates that disrupt protein binding. This kit assesses compound-based disruption of Bcl2L1-Bax binding via detection of Bax levels. This product uses EFC technology. The cell line included overexpresses Bcl2L1 target in an untagged form and Bax binding partner fused to PL. When a compound engages the Bcl2L1 target protein, partner binding is disrupted and the destabilized Bax is degraded, leading to a loss of EFC signal upon addition of EA containing detection reagent. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates.

Specifications

Assay Entry U2OS
For Use With (Application) Assay-Ready eXpress Kits
Format 1,000 dp (10 x 96-well)
Primary Agonist ABT-737
Quantity 1,000 dp (10 x 96-well)
Target Entry Bcl2L1-Bax
Cell Line U2OS
Target Bcl2L1-Bax
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.