missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX InCELL Hunter™ eXpress Bcl2L1-Bax Protein Binding Assay
Click to view available options
Quantity:
200 dp (2 x 96-well)
Unit Size:
1 set
Description
The InCELL Hunter™ eXpress Bcl2L1-Bax Protein Binding Assay provides an easy-to-use cell based assay to study small molecule drug candidates that disrupt protein binding. This kit assesses compound-based disruption of Bcl2L1-Bax binding via detection of Bax levels. This product uses EFC technology. The cell line included overexpresses Bcl2L1 target in an untagged form and Bax binding partner fused to PL. When a compound engages the Bcl2L1 target protein, partner binding is disrupted and the destabilized Bax is degraded, leading to a loss of EFC signal upon addition of EA containing detection reagent. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates.
Specifications
Specifications
| Assay Entry | U2OS |
| For Use With (Application) | Assay-Ready eXpress Kits |
| Primary Agonist | ABT-737 |
| Quantity | 200 dp (2 x 96-well) |
| Target Entry | Bcl2L1-Bax |
| Cell Line | U2OS |
| Target | Bcl2L1-Bax |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction