Learn More
Novus Biologicals™ ImmPACT (TM) DAB HRP Substrate
Product Code. 18626410 Shop All Bio Techne Products
Click to view available options
Quantity:
120 mL
Unit Size:
120mL
Product Code. 18626410
Brand: Novus Biologicals™ SK4105NB
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Specifications
For Use With (Application) This product is an entirely new formula for a diaminobenzidine based peroxidase substrate. ImmPACT(TM) DAB produces a dark brown reaction product that is crisper and is 3-4 times more sensitive than the original Vector DAB substrate kit. It can be used for both manual and automated staining methods. Sections stained with DAB can also be viewed by darkfield and electron microscopy. Slides developed with DAB can be dehydrated, cleared, and permanently mounted. The ImmPACT(TM) DAB working solution is stable for at least five days at room temperature and at least two weeks when stored at 4°C. This product consists of 120 mL of diluent and a convenient concentrated stock solution of highly purified DAB. This product can also be used on blots.
Quantity 120 mL
Substrate Type DAB HRP Substrate
Storage Requirements Store at 4°C in the dark.