missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ ImmPACT (TM) DAB EqV HRP Substrate
Shop All Bio Techne Products
Click to view available options
Quantity:
100 mL
400 mL
Unit Size:
100mL
400mL
Specifications
Specifications
| Quantity | 100 mL |
| Substrate Type | DAB Substrate |
| Item Description | ImmPACT(TM) DAB EqV (equal volume) consists of 200 ml each of Reagent 1 and Reagent 2 which when mixed together in equal volumes form the working solution. ImmPACT(TM) DAB EqV produces a dark brown reaction product that is 3 to 5 times more sensitive than the original DAB substrate. The working solution is stable for several days when stored at 4°C. This substrate can be used for both manual and automated staining methods. ImmPACT(TM) DAB EqV can be used for blot detection on nylon and PVDF membranes but is not recommended for nitrocellulose membranes. Sections stained with DAB can also be viewed by darkfield and electron microscopy. Slides developed with DAB can be dehydrated, cleared, and permanently mounted. |
| Storage Requirements | Store at 4°C in the dark. |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction