missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ ImmPACT (TM) DAB EqV HRP Substrate

Product Code. 18334314 Shop All Bio Techne Products
Click to view available options
Quantity:
100 mL
400 mL
Unit Size:
100mL
400mL
This item is not returnable. View return policy

Product Code. 18334314

Brand: Novus Biologicals™ SK4103NB100ML

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

Optimized for IHC/ICC/ISH

TRUSTED_SUSTAINABILITY

Specifications

Quantity 100 mL
Substrate Type DAB Substrate
Item Description ImmPACT(TM) DAB EqV (equal volume) consists of 200 ml each of Reagent 1 and Reagent 2 which when mixed together in equal volumes form the working solution. ImmPACT(TM) DAB EqV produces a dark brown reaction product that is 3 to 5 times more sensitive than the original DAB substrate. The working solution is stable for several days when stored at 4°C. This substrate can be used for both manual and automated staining methods. ImmPACT(TM) DAB EqV can be used for blot detection on nylon and PVDF membranes but is not recommended for nitrocellulose membranes. Sections stained with DAB can also be viewed by darkfield and electron microscopy. Slides developed with DAB can be dehydrated, cleared, and permanently mounted.
Storage Requirements Store at 4°C in the dark.

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.