Learn More
Merck Immobilon NOW Dispenser
Product Code. 16389591 Shop All Merck Products
Change view
Click to view available options
Quantity:
1
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16389591 1 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16389591 Supplier Merck Supplier No. IMDISP
Only null left
Add to basket
Add to basket
Includes
Immobilon NOW Dispenser
Immobilon Transfer Membranes offer the convenience of pre-cut sheets with the flexibility of membrane rolls
  • Single-cut convenience
  • Compatibility with mini or midi gels
  • No waste
  • Compact package saves valuable lab space
TRUSTED_SUSTAINABILITY
Specifications
Description Membrane dispenser
For Use With (Application) Immunoblotting, Dot blotting
Quantity 1