Learn More
Cytiva Immobiline™ DryStrip Kit and Components
Product Code. 10239624 Shop All Cytiva Products
Click to view available options
Type:
Anode
Cathode
Dry Strip Slinger
Immobiline DryStrip Kit
Sample Cup
Sample Cup Bar
Tray and Electrode Holder
Unit Size:
1 set
Each
Pack of 4
Pack of 60
Product Code. 10239624
Brand: Cytiva 18100434
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Precast Immobiline DryStrip gels for running the first dimension of a 2-D electrophoresis experiment with the Multiphor II electrophoresis system
Run up to 12 Immobiline DryStrip strips simultaneously with up to 100μL on each Immobiline DryStrip.
  • The instruction manual presents all necessary information for the entire method for horizontal 2-D electrophoresis. By appropriate adaptations, the SDS electrophoresis can also be performed using vertical electrophoresis equipment
  • Immobiline DryStrip kit facilitates sample application and running and equilibration of Immobiline DryStrip for the first dimension in 2-D (two-dimensional) electrophoresis
  • Isoelectric focusing with an immobilized pH gradient (IPG), makes true isoelectric focusing possible and significantly improves the reproducibility of the spot distribution along the pH gradient axis of 2-D maps
  • Individual components are also available separately
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Multiphor II Electrophoresis System
Type Dry Strip Slinger