Learn More
Ebro Electronic™ Thermocouple Rod Probes
Product Code. 11703489
Click to view available options
Length (Metric) Probe:
185 mm
300 mm
Diameter (Metric) Probe:
0.5 mm
1.5 mm
3 mm
Model:
TPN 100
TPN 120
TPN 120-30
TPN 140
TPN 140-30
Unit Size:
Each
Product Code. 11703489
Brand: Ebro Electronic™ 13410611
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Rod probes for temperature measurement of air, ovens, fluid and gases.
For various applications in food, laboratory, industial use.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance RoHS Directive 2011/65/EU
Diameter (Metric) Probe 0.5 mm
Disposable Single-use
For Use With (Application) Measurement of air, oven, fluids and gas temperatures
Height (Metric) 15 mm
Immersion Type Total or Partial
Length (Metric) 220 mm
Length (Metric) Probe 185 mm
Material Metal
Measurement Time 0.4 sec.
Model TPN 100
Probe Type NiCrNi rod
Weight (Metric) 0.04 kg
Product Type Probe
Temperature Range (English) -40 °F to +2012 °F
Temperature Range (Metric) -40°C to +1100°C
For Use With (Equipment) TFN 520
Width (Metric) 15 mm