Learn More
Fisherbrand™ Immersion Basket with Handle
Product Code. 11786198 Shop All Fisherbrand Products
Click to view available options
For Use With (Equipment):
Ultrasonic Bath, Elmasonic X-tra ST 1400H
Ultrasonic Bath, Elmasonic Xtra Basic 1600 160 L
Ultrasonic Bath, Elmasonic Xtra Basic 2500 250 L
Ultrasonic Bath, Elmasonic Xtra Basic 300 30 L
Ultrasonic Bath, Elmasonic Xtra Basic 550 55L
Ultrasonic Bath, Elmasonic Xtra Basic 800 80 L
Ultrasonic Cleaner, Elmasonic S450 H 45 L
missing translation for 'unitSize'
Each
Product Code. 11786198
Only null left
Add to basket
Add to basket
Facilitates complete immersion within S-line Ultrasonic Cleaner baths. Fisherbrand™ Immersion Baskets with Handle are manufactured from stainless steel.
Designed for the complete immersion of items and parts in Fisherbrand™ 112xxx Series Dual Frequency Ultrasonic Cleaners. Choose the Fisherbrand™ Stainless Steel Immersion Basket with Handles designed for the specific ultrasonic cleaner being used.
  • Mesh bottom
  • One handle on each end
  • Functional and modern design
  • Available for a variety of capacities
Specifications
Type Basket
For Use With (Equipment) Ultrasonic Cleaner, Elmasonic S450 H 45 L
Material Stainless Steel