Learn More
Cytiva Solution dNTPs
Product Code. 10527065 Shop All Cytiva Products
Change view
Click to view available options
Product Type:
dATP
dCTP
dGTP
dTTP
dUTP
Quantity:
25 μm
100 μm
500 μm
Unit Size:
Each
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Quantity Unit Size
10527065 dTTP 100 μm Each
10238084 dATP 25 μm Each
10545335 dGTP 25 μm Each
10389764 dGTP 100 μm Each
10648035 dCTP 25 μm Each
10430125 dCTP 100 μm Each
10074294 dTTP 25 μm Each
10024684 dUTP 25 μm Each
10526875 dGTP 500 μm Each
10004824 dATP 100 μm Each
10665355 dUTP 100 μm Each
10537065 dTTP 500 μm Each
10350055 dATP 500 μm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Product Code. 10527065 Supplier Cytiva Supplier No. 28406532
  • ONLINE EXCLUSIVE PRICE
    170.00€
    155.36€ / Pack of 1
    Save 14.64€
    8% Off
 Restricted Item
In Stock
Use dNTPs incorporating the most recent advances in manufacturing technology. Cytiva illustra™ Solution dNTPs offer the highest purity deoxynucleotides available for amplification, dideoxy sequencing, labeling, mutagenesis, cDNA synthesis, and expression profiling.
  • Free from DNase, RNase, and nicking enzyme activity
  • Greater than 99% triphosphate purity (by HPLC) for optimum performance and consistency
  • Buffer-free and ready-to-use solutions in multiple formats
  • Functionally tested for long PCR and sequencing
Order Info
Custom formulation, testing, packaging, and labeling are available
TRUSTED_SUSTAINABILITY
Specifications
Format Liquid
pH 7
Product Type dTTP
Concentration 100 mM
Purity >99%
Quantity 100 μm
Color Colorless
For Use With (Application) Functionally tested for long PCR and sequencing, Nucleic Acid amplification, labelling, mutagenesis, cDNA synthesis and expression profiling