Learn More
Zeiss™ Coaxial Epi-Illumination S
Product Code. 11895470
Change view
Click to view available options
Type:
Illuminator
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
11895470 Illuminator Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11895470 Supplier Zeiss™ Supplier No. 4351120000000
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
Mountable at mount S with diam. 76mm support (435402). Zeiss Coaxial Epi-Illumination S is recommended for objective Plan Apo S 1.0x (435203), further combinations of objectives S ≥1.0x and eyepieces possible.
Compatible Microscopes:
  • SteREO Discovery.V12
  • SteREO Discovery.V20
  • SteREO Discovery.V8
  • SteREO Lumar.V12
Specifications
Type Illuminator
Description Mountable at mount S with diam. 76mm support
For Use With (Equipment) Objective Plan Apo S 1.0x
Quantity 1 Ea.