Learn More
Abnova™ IL22RA1 Recombinant Protein
Human IL22RA1 full-length ORF recombinant protein with GST-tag at N-terminal
Brand: Abnova™ H00058985-P01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene belongs to the class II cytokine receptor family, and has been shown to be a receptor for interleukin 22 (IL22). IL22 receptor is a protein complex that consists of this protein and interleukin 10 receptor, β (IL10BR/CRFB4), a subunit also shared by the receptor complex for interleukin 10 (IL10). This gene and interleukin 28 receptor, α (IL28RA) form a cytokine receptor gene cluster in the chromosomal region 1p36.
- Theoretical MW (kDa): 86.9
- Preparation method: In vitro wheat germ expression system
- Purification: Glutathione Sepharose 4 fast flow
- Storage buffer: 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer
Sequence: EDPSDLLQHVKFQSSNFENILTWDSGPEGTPDTVYSIEYKTYGERDWVAKKGCQRITRKSCNLTVETGNLTELYYARVTAVSAGGRSATKMTDRFSSLQHTTLKPPDVTCISKVRSIQMIVHPTPTPIRAGDGHRLTLEDIFHDLFYHLELQVNRTYQMHLGGKQREYEFFGLTPDTEFLGTIMICVPTWAKESAPYMCRVKTLPDRTWTYSFSGAFLFSMGFLVAVLCYLSYRYVTKPPAPPNSLNVQRVLTFQPLRFIQEHVLIPVFDLSGPSSLAQPVQYSQIRVSGPREPAGAPQRHSLSEITYLGQPDISILQPSNVPPPQILSPLSYAPNAAPEVGPPSYAPQVTPEAQFPFYAPQAISKVQPSSYAPQATPDSWPPSYGVCMEGSGKDSPTGTLSSPKHLRPKGQLQKEPPAGSCMLGGLSLQEVTSLAMEESQEAKSLHQPLGICTDRTSDPNVLHSGEEGTPQYLKGQLPLLSSVQIEGHPMSLPLQPPSRPCSPSDQGPSPWGLLESLVCPKDEAKSPAPETSDLEQPTELDSLFRGLALTVQWES
Best use within three months from the date of receipt of this protein.
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
AAH29273 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
87.3 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
EDPSDLLQHVKFQSSNFENILTWDSGPEGTPDTVYSIEYKTYGERDWVAKKGCQRITRKSCNLTVETGNLTELYYARVTAVSAGGRSATKMTDRFSSLQHTTLKPPDVTCISKVRSIQMIVHPTPTPIRAGDGHRLTLEDIFHDLFYHLELQVNRTYQMHLGGKQREYEFFGLTPDTEFLGTIMICVPTWAKESAPYMCRVKTLPDRTWTYSFSGAFLFSMGFLVAVLCYLSYRYVTKPPAPPNSLNVQRVLTFQPLRFIQEHVLIPVFDLSGPSSLAQPVQYSQIRVSGPREPAGAPQRHSLSEITYLGQPDISILQPSNVPPPQILSPLSYAPNAAPEVGPPSYAPQVTPEAQFPFYAPQAISKVQPSSYAPQATPDSWPPSYGVCMEGSGKDSPTGTLSSPKHLRPKGQLQKEPPAGSCMLGGLSLQEVTSLAMEESQEAKSLHQPLGICTDRTSDPNVLHSGEEGTPQYLKGQLPLLSSVQIEGHPMSLPLQPPSRPCSPSDQGPSPWGLLESLVCPKDEAKSPAPETSDLEQPTELDSLFRGLALTVQWES | |
CRF2-9/IL22R/IL22R1 | |
IL22RA1 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Protein Array, ELISA, Western Blot | |
58985 | |
IL22RA1 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
IL22RA1 | |
Human | |
Recombinant | |
Solution |