missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ IL13RA1 (Human) Recombinant Protein
Human IL13RA1 full-length ORF ( AAH15768, 1 a.a. - 427 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00003597-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MECPARLCGLWALLLCAGGGGGSGGAAPTETQPPVTNLSVSVENLCTVIWTWNPPEGASSNCSLWYFSHFGDKQDKKIAPETRRSIEVPLNERICLQVGSQCSTNESEKPSILVEKCISPPEGDPESAVTELQCIWHNLSYMKCSWLPGRNTSPDTNYTLYYWHRSLEKIHQCENIFREGQYFGCSFDLTKVKDSSFEQHSVQIMVKDNAGKIKPSFNIVPLTSRVKPDPPHIKNLSFHNDDLYVQWENPQNFISRCLFYEVEVNNSQTETHNVFYVQEAKCENPEFERNVENTSCFMVPGVLPDTLNTVRIRVKTNKLCYEDDKLWSNWSQEMSIGKKRNSTLYITMLLIVPVIVAGAIIVLLLYLKRLKIIIFPPIPDPGKIFKEMFGDQNDDTLHWKKYDIYEKQTKEETDSVVLIENLKKASQ
Specifications
AAH15768 | |
interleukin 13 receptor, alpha 1 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
IL13RA1 | |
GST |
3597 | |
Wheat germ expression system | |
25 μg | |
CD213A1, IL-13Ra, NR4 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |