missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ IL13 (Human) Recombinant Protein
Human IL13 full-length ORF ( NP_002179, 1 a.a. - 132 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00003596-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MALLLTTVIALTCLGGFASPGPVPPSTALRELIEELVNITQNQKAPLCNGSMVWSINLTAGMYCAALESLINVSGCSAIEKTQRMLSGFCPHKVSAGQFSSLHVRDTKIEVAQFVKDLLLHLKKLFREGRFN
Specifications
NP_002179 | |
interleukin 13 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
IL13 | |
GST |
3596 | |
Wheat germ expression system | |
25 μg | |
ALRH, BHR1, IL-13, MGC116786, MGC116788, MGC116789, P600 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |