missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-27R alpha/WSX-1/TCCR Antibody (MM0563-8A5), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-11917AF647
Additional Details : Weight : 0.00970kg
Description
IL-27R alpha/WSX-1/TCCR Monoclonal specifically detects IL-27R alpha/WSX-1/TCCR in Human samples. It is validated for Western Blot.Specifications
IL-27R alpha/WSX-1/TCCR | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
IL27RA | |
Human recombinant IL-27R alpha/WSX-1/TCCR (extracellular domain) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
MM0563-8A5 | |
Western Blot | |
class I cytokine receptor, CRL1IL-27R, Cytokine receptor-like 1, IL27R, IL-27RA, IL-27R-alpha, interleukin 27 receptor, alpha, interleukin-27 receptor subunit alpha, TCCRIL-27 receptor subunit alpha, T-cell cytokine receptor type 1, Type I T-cell cytokine receptor, WSX-1, WSX1IL-27R subunit alpha, zcytor1 | |
Mouse | |
Protein G purified | |
Apoptosis, Cytokine Research | |
9466 | |
Store at 4C in the dark. | |
IgG2 |