missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ IKBKB (Human) Recombinant Protein
Human IKBKB full-length ORF ( AAH06231.1, 1 a.a. - 256 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00003551-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MSWSPSLTTQTCGAWEMKERLGTGGFGNVIRWHNQETGEQIAIKQCRQELSPRNRERWCLEIQIMRRLTHPNVVAARDVPEGMQNLAPNDLPLLAMEYCQGGDLRKYLNQFENCCGLREGAILTLLSDIASALRYLHENRIIHRDLKPENIVLQQGEQRLIHKIIDLGYAKELDQGSLCTSFVGTLQYLAPELLEQQKYTVTVDYWSFGTLAFECITGFRPFLPNWQPVQCVRMWPGTVAHSCNPSTLGGRGRWIS
Specifications
AAH06231.1 | |
inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase beta | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
IKBKB | |
GST |
3551 | |
Wheat germ expression system | |
25 μg | |
FLJ40509, IKK-beta, IKK2, IKKB, MGC131801, NFKBIKB | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |