missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ IkB-alpha Antibody Pair [HRP]

Product Code. 18272497 Shop All Bio Techne Products
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
This item is not returnable. View return policy

Product Code. 18272497

Brand: Novus Biologicals™ NBP27938315PLATES

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human IkB-alpha.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 12.5 pg/mL
Assay Sensitivity 12.5-800 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human IkB-alpha.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias alpha, I-kappa-B-alpha, IkappaBalphaNF-kappa-B inhibitor alpha, IKBAikB-alpha, IkB-alpha, MAD3, MAD-3, Major histocompatibility complex enhancer-binding protein MAD3, NFKBI, nuclear factor of kappa light chain gene enhancer in B-cells, nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor
Gene ID (Entrez) 4792
Gene Symbol NFKBIA
Host Species Mouse/Rabbit
Kit Contents
  • Detection Antibody: 0.5 mg/mL mouse anti-IkB-alpha monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % glycerol, pH 7.4).
  • Capture Antibody: 1.0 mg/mL of rabbit anti-IkB-alpha monoclonal antibody (in PBS, pH 7.4).
  • Standard: Each vial contains 22 ng of recombinant IkB-alpha.
  • Quantity 15 Plates
    Regulatory Status RUO
    Research Discipline Apoptosis, Cell Biology, Chaperone Mediated Autophagy (CMA), Diabetes Research, DNA Repair, Nucleotide Excision Repair, Phospho Specific, Signal Transduction
    Storage Requirements Storage is content dependent.
    Target IkB-alpha
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 0.8 ng/mL is recommended.
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.