Learn More
Proteintech IHCeasy NOLC1 Ready-To-Use IHC Kit
Product Code. 18025505 Shop All Proteintech Products
Click to view available options
Quantity:
50 Tests
Unit Size:
50 tests
Product Code. 18025505
Brand: Proteintech KHC174350T
KHC1743 is a ready-to-use IHC kit for staining of NOLC1.
KHC1743 is a ready-to-use IHC kit for staining of NOLC1. The kit provides all reagents, from antigen retrieval to cover slip mounting, that require little to no diluting or handling prior to use. Simply apply the reagents to your sample slide according to the protocol and you're steps away from obtaining high-quality IHC data.
Specifications
Quantity 50 Tests
Form Liquid