Learn More
Proteintech IHCeasy HIST1H3A Ready-To-Use IHC Kit
Product Code. 18065485 Shop All Proteintech Products
Click to view available options
Quantity:
50 Tests
Unit Size:
50 tests
Product Code. 18065485
Brand: Proteintech KHC167550T
KHC1675 is a ready-to-use IHC kit for staining of HIST1H3A.
KHC1675 is a ready-to-use IHC kit for staining of HIST1H3A. The kit provides all reagents, from antigen retrieval to cover slip mounting, that require little to no diluting or handling prior to use. Simply apply the reagents to your sample slide according to the protocol and you're steps away from obtaining high-quality IHC data.
Specifications
Quantity 50 Tests
Form Liquid