missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ IGJ (Human) Recombinant Protein
Human IGJ full-length ORF ( AAH38982, 1 a.a. - 159 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00003512-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MKNHLLFWGVLAVFIKAVHVKAQEDERIVLVDNKCKCARITSRIIRSSEDPNEDIVERNIRIIVPLNNRENISDPTSPLRTRFVYHLSDLCKKCDPTEVELDNQIVTATQSNICDEDSATETCYTYDRNKCYTAVVPLVYGGETKMVETALTPDACYPD
Specifications
AAH38982 | |
immunoglobulin J polypeptide, linker protein for immunoglobulin alpha and mu polypeptides | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
IGJ | |
GST |
3512 | |
Wheat germ expression system | |
25 μg | |
IGCJ, JCH | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |