missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ IGFBP4 (Human) Recombinant Protein
Human IGFBP4 full-length ORF ( AAH16041, 1 a.a. - 258 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00003487-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MLPLCLVAALLLAAGPGPSLGDEAIHCPPCSEEKLARCRPPVGCEELVREPGCGCCATCALGLGMPCGVYTPRCGSGLRCYPPRGVEKPLHTLMHGQGVCMELAEIEAIQESLQPSDKDEGDHPNNSFSPCSAHDRRCLQKHFAKIRDRSTSGGKMKVNGAPREDARPVPQGSCQSELHRALERLAASQSRTHEDLYIIPIPNCDRNGNFHPKQCHPALDGQRGKCWCVDRKTGVKLPGGLEPKGELDCHQLADSFRE
Specifications
AAH16041 | |
insulin-like growth factor binding protein 4 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
IGFBP4 | |
GST |
3487 | |
Wheat germ expression system | |
25 μg | |
BP-4, HT29-IGFBP, IBP4, IGFBP-4 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |