Learn More
IFN-gamma R1/CD119 Mouse anti-Human, Alexa Fluor 350, Clone: 92101, R&D Systems™
Mouse Monoclonal Antibody
Brand:  R&D Systems FAB6731U-100UG
Encompass
Product Code. 15778313
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
IFN-gamma R1/CD119 Monoclonal specifically detects IFN-gamma R1/CD119 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
IFN-gamma R1/CD119
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
P15260.1
IFNGR1
Mouse myeloma cell line NS0-derived recombinant human IFN-gamma R1 Glu18-Gly245 Accession # P15260.1
RUO
3459
Human
IgG1
Flow Cytometry
92101
Alexa Fluor 350
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
AVP, type 2, CD119, CD119 antigen, FLJ45734, IFN-gamma receptor 1, IFNgammaR1, IFN-gamma-R1, IFNGR, IFN-gR1, IFNGR1, immune interferon receptor 1, interferon gamma receptor 1, interferon-gamma receptor alpha chain, type 2
Mouse
100 μg
Primary
Detects human IFN-gamma R1 in direct ELISAs and Western blots. In direct ELISAs, this antibody does not cross-react with recombinant mouse IFN-gamma R1, recombinant human (rh) IFN-gamma R2, or rhIL-10 R beta .
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Title
Select an issue
For Research Use Only (RUO)