missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFN-epsilon Mouse anti-Human, Clone: 983338, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB9147-100
This item is not returnable.
View return policy
Description
IFN-epsilon Monoclonal specifically detects IFN-epsilon in Human samples. It is validated for ELISA, Immunohistochemistry.Specifications
| IFN-epsilon | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| IFNE, IFN-E, IFNE1PRO655, IFNepsilon, IFN-epsilon, IFNT1, Ifn-tau-11, INFE1, interferon epsilon, interferon epsilon 1, Interferon epsilon-1, interferon tau-1, interferon, epsilon, interferon-epsilon, MGC119018, MGC119020 | |
| E. coli-derived recombinant human IFN-epsilon Leu22-Arg208 Accession # Q86WN2 | |
| 100 μg | |
| 338376 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Immunohistochemistry | |
| 983338 | |
| Immunohistochemistry 1-25 ug/mL | |
| Q86WN2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human IFN-epsilon in direct ELISAs. | |
| Human | |
| Supernatant |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction