missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ IEM 1460
GREENER_CHOICE

Product Code. 18025922
Click to view available options
Quantity:
10 mg
50 mg
missing translation for 'unitSize'
10mg
50mg
This item is not returnable. View return policy

Product Code. 18025922

missing translation for 'mfr': Tocris Bioscience™ 1636/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Open-channel blocker of AMPA currents; selective for non-GluA2-containing receptors

Voltage-dependent open-channel blocker of AMPA receptors. Selective for GluR2 subunit-lacking (Ca2+-permeable) receptors over GluA2-containing receptors (IC50 values are 2.6 and 1102 μM, respectively). Selectively blocks fast spiking interneuron but not medium spiny projection neuron in mouse striatum. Also blocks NMDA receptor-mediated currents. Anticonvulsant in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121034-89-7
Molecular Formula C19H38Br2N2

Specifications

Quantity 50 mg
Formula Weight 454.33
Purity >98%
Molecular Weight (g/mol) 454.335
Chemical Name or Material IEM 1460
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.