Learn More
Thermo Scientific™ IEF Sample Buffer, 4X, pH 3-7 with lysine
Product Code. 15415819
Change view
Click to view available options
Quantity:
25 mL
50 mL
Unit Size:
25mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15415819 25 mL 25mL
15425819 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15415819 Supplier Thermo Scientific™ Supplier No. J63633.AC
Request Bulk or Custom Format
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Concentration 4X
pH Range 3 to 7
Form Liquid
Quantity 25 mL
Product Type Sample Loading Buffer