Learn More
Serva Electrophoresis™ IEF Sample Buffer, 2X
Product Code. 11802140
Encompass_Preferred
Change view
Click to view available options
Quantity:
20mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11802140 20mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11802140 Supplier Serva Electrophoresis™ Supplier No. 4253701
 Restricted Item
Estimated Shipment: 29-07-2026
Specifications
Color Red
Concentration 2X
pH Range 6.5 to 7.5
Form Solution
Material 4% Servalyt™ 4 to 9 T, 30% glycerol and 0.005% phenol red
Odor Odorless
Quantity 20mL
Sterility Sterile
For Use With (Equipment) Vertical and Horizontal IEF Applications and Systems