missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ ICOSLG Recombinant Protein

Brand:  Abnova™ H00023308-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16157152

  • 508.00€ / 25µg
Estimated Shipment: 07-06-2024
to see stock.
Add to basket

Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH64637
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
57.2
8
Glutathione Sepharose 4 Fast Flow
25 ug
TQEKEVRAMVGSDVELSCACPEGSRFDLNDVYVYWQTSESKTVVTYHIPQNSSLENVDSRYRNRALMSPAGMLRGDFSLRLFNVTPQDEQKFHCLVLSQSLGFQEVLSVEVTLHVAANFSVPVVSAPHSPSQDELTFTCTSINGYPRPNVYWINKTDNSLLDQALQNDTVFLNMRGLYDVVSVLRIARTPSVNIGCCIENVLLQQNLTVGSQTGNDIGERDKITENPVSTGEKNAATWSILAVLCLLVVVAVAIGWVCRDRCLQHSYAGAWAVSPETELTGHV
B7-H2/B7H2/B7RP-1/B7RP1/CD275/GL50/ICOS-L/ICOSL/KIAA0653/LICOS
ICOSLG
Wheat Germ (in vitro)
GST
Antibody Production, Protein Array, ELISA, Western Blot
23308
ICOSLG (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ICOSLG
Human
Recombinant
Solution
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers