missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ ICG-d7
GREENER_CHOICE

Product Code. 30000017
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30000017 10 mg 10mg
30000019 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30000017 Supplier Tocris Bioscience™ Supplier No. 7749/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Near-infrared fluorescent dye; partially deuterated form of Indocyanine Green (Cat. No. 7510)

ICG-d7 is a partially deuterated form of near-infrared fluorescent dye Indocyanine Green (ICG; Cat. No. 7510). Operates in the 700 - 900 nm wavelength range; also exhibits fluorescence emission in the shortwave infrared window (SWIR; 1,000 - 2,000 nm) which enables in vivo imaging. ICG-d7 shows improved stability in aqueous solutions over ICG with the same fluorescence brightness, and similar absorption and emission maxima in both DMSO and FBS. Excitation maximum = 794 nm; emission maximum = 818 nm.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium 4-(2-((1E,3E,5E,7Z)-7-(1,1-Dimethyl-3-(4-sulfonatobutyl)-1,3-dihydro-2H-benzo[e]indol-2-ylidene)hepta-1,3,5-trien-1-yl-1,2,3,4,5,6,7-d7)-1,1-dimethyl-1H-benzo[e]indol-3-ium-3-yl)butane-1-sulfonate
CAS 2861905-57-7
Quantity 10 mg
Target Fluorescent Dyes
Molecular Formula C43H40D7N2O6S2Na
Purity 0.95
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.