missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ ICG 001

Product Code. 18065922
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18065922 10 mg 10mg
18035112 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18065922 Supplier Tocris Bioscience™ Supplier No. 4505/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Inhibits TCF/β-catenin-mediated transcription

ICG 001 is an inhibitor of TCF/β-catenin-mediated transcription. Selectively inhibits β-catenin/CBP interaction; displays no effect on β-catenin/p300 interaction. Exhibits growth inhibitory effects in colon carcinoma cell lines (SW480 and HCT-116 cells); also displays efficacy in Min mouse and nude mouse SW620 xenograft models.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (6S,9aS)-Hexahydro-6-[(4-hydroxyphenyl)methyl]-8-(1-naphthalenylmethyl)-4,7-dioxo-N-(phenylmethyl)-2H-pyrazino[1,2-a]pyrimidine-1(6H)-carboxamide
CAS 780757-88-2
Quantity 10 mg
Target beta-Catenin Inhibitors
Molecular Formula C33H32N4O4
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.