Learn More
Abnova™ ICAM1 (Human) Recombinant Protein (P01)
Human ICAM1 full-length ORF with GST-tag at N-terminal
Brand: Abnova™ H00003383-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a cell surface glycoprotein which is typically expressed on endothelial cells and cells of the immune system. It binds to integrins of type CD11a / CD18, or CD11b / CD18 and is also exploited by Rhinovirus as a receptor.
- Theoretical MW: 84.26kDa
- Preparation Method: In vitro wheat germ expression system
- Purification: Glutathione Sepharose 4 Fast Flow
- Storage Buffer: 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
Antibody Production, ELISA, Protein Array, Western Blotting (Recombinant Protein) | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
Human ICAM1 Full-length ORF Recombinant Protein with GST-tag at N-terminal | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE stained with Coomassie Blue | |
Wheat Germ (in vitro) | |
Human |
Solution | |
84.26 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
25μg | |
-80°C | |
Yes |
For Research Use Only