Learn More
Brady™ i75 Series R4900 Ribbon for the i7500 Label Printer
Product Code. 30279153
Click to view available options
Alternate Part Number:
i75-R4900
i75-R4902
Unit Size:
Each
Product Code. 30279153
Brand: Brady™ 176750
Only null left
Add to basket
Add to basket
i75-Series ribbon with LabelSense™ Technology warns user if ribbon is incompatible with installed i75-Series label material.
  • i75-series ribbon is self-centering with ink-side always correctly facing the label
  • One-color ribbon for printing a single color onto a label
  • R4900 Series Resin formulation offers excellent resistance to solvents, smears, scratches and high heat when printed on the appropriate label material from Brady™
  • Combine Brady label materials with their recommended ribbon series for superior legend performance
Specifications
Product Type Printer Ribbon
Color Black
For Use With (Equipment) i7500
Material Polyester
Print Technology Ink Jet
Length (English) 328.08 yd.
Width (English) 2.36 in.
Length (Metric) 300 m
Width (Metric) 60 mm
Thickness (Metric) 4.5 μm
Dimensions (L x W) 60 mm x 300 m
Quantity 1 Ea.
Alternate Part Number i75-R4902