Learn More
Brady™ i3300 Industrial Label Printer Kit EU with Wi-Fi and Workstation SFID Suite
Product Code. 16346925
Change view
Click to view available options
:
i3300 Wi-Fi Kit EU with SFID; Color Capability: Monochrome; Print Speed: 101.6 mm/s; Print Technology: Thermal Transfer; Resolution: 300 dpi; Weight (Metric): 5.2 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16346925 i3300 Wi-Fi Kit EU with SFID; Color Capability: Monochrome; Print Speed: 101.6 mm/s; Print Technology: Thermal Transfer; Resolution: 300 dpi; Weight (Metric): 5.2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16346925 Supplier Brady™ Supplier No. 241119
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
The BradyPrinter i3300 Industrial Label Printer can print a wide range of sign and labels in monocolor, including labels to identify wires, laboratory samples, and facilities without any hassle.
This mid-volume, i3300 Industrial Label Printer is ideal for identification throughout your facility, including everything from wires and panels to safety and facility labels.
  • Wi-Fi connectivity allows you to get data more easily from a variety of sources
  • Print smart technology allows printing on the first label- even sleeves and small labels
  • Foolproof drop-in label rolls and ribbons snap into place in 20 seconds
  • Prints on more than 825 parts and 70 material options for facility-wide identification
  • Contact our Brady™ Technical Support Team to schedule a free phone appointment
  • Includes EU power cord and Brady™ Workstation Safety & Facility Identification Suite
Specifications
Display Type Color LCD Touchscreen
Print Technology Thermal Transfer
Width (English) 9 in.
Width (Metric) 229 mm
Height (English) 9.5 in.
Height (Metric) 241 mm
Depth (English) 12 in.
Depth (Metric) 305 mm
Autoclavable Not Autoclavable
Color Capability Monochrome
Includes i3300 Printer, EU Power Cord, Brady™ Workstation SFID Suite, USB Cable, Product CD, Cleaning Kit, Printed Quick Start Guide
Languages Bulgarian, Chinese (Tradditional), Chinese (Simplified), Croatian, Czech, Danish, Dutch, English, Estonian, Finnish, French, German, Hungarian, Italian, Norwegian, Polish, Portuguese, Romanian, Russian, Spanish, Swedish, Turkish
Memory 195 MB
Model i3300
Print Speed 101.6 mm/s
Resolution 300 dpi
Connector Types Ethernet, USB, USB 2.0
Operating System Windows™ 10, Windows™ 7, Windows™ 8, Windows™ Servers 2012, 2012 R2, and 2016
Temperature Range (Metric) 10°C to 40°C
Weight (English) 11.5 lb.
Weight (Metric) 5.2 kg
For Use With (Application) Barcode and Inventory Labeling, Hazard Identification, Laboratory Labeling
Warranty 1 Year
Voltage 110/240 V
Alternate Part Number i3300-C-EU-W-SFIDS