Learn More
Cytiva E. coli BL21
Product Code. 10658225 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 Vial
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10658225 1 Vial Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10658225 Supplier Cytiva Supplier No. 27154201
 Restricted Item
Estimated Shipment: 19-11-2026
Protease-deficient E. coli host strain for optimal expression of recombinant proteins
  • F-, ompT, hsdS (rB-, mB-), gal, dcm
  • Exemplary host well suited for use with the GST gene fusion system
  • High-level expression of a variety of recombinant proteins
  • Defective in E. coli strain B, and wild-type restriction systems to permit high efficiency transformation with unmodified plasmid DNA
  • Verified by RAPD fingerprint analysis using Ready-To-Go™ RAPD analysis Kit
  • Lacks the major Lon protease responsible for degradation of damaged or foreign proteins
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Vial
Species E. coli