Learn More
STEMCELL Technologies Hypoxia Incubator Chamber
Description
- The Hypoxia Incubator Chamber is a self-contained and sealed chamber that fits inside existing laboratory incubators
- Each unit utilizes a surface-type seal in which all portions of the O-ring are uniformly compressed by a ring clamp for a reliable, air-tight seal
- The cylindrical walls and semi-spherical top and bottom minimize gas flow resistance and eliminate dead space during initial purging
- The chambers have an integrated stacking feature for storage during or after experimentation
- All units are molded from high quality polycarbonate and will not break, crack or cloud with extended use
- Capacity: 84 x 35 mm plates, 27 x 60 mm plates, 12 x 100 mm plates, 12 x 96-well plates, 18 x 25 cm2 flasks
- Hypoxia Incubator Chamber (Catalog #27310) Components: Lid* (Polycarbonate)
- Base* (Polycarbonate) with O-ring and Tubing Clamp
- Tray* (Polycarbonate)
- Ring Clamp
- O-ring
- Tubing Clamp with Tubing
Specifications
Specifications
| For Use With (Application) | Cell Culture |
| Includes | 1 Chamber |