Learn More
STEMCELL Technologies HypoThermosol™ FRS
Animal component-free, defined hypothermic (2 - 8°C) preservation medium for a range of cell and tissue types
Brand: STEMCELL Technologies 07934
Additional Details : Weight : 0.16000kg
Description
- HypoThermosol™ FRS is an optimized hypothermic (2 - 8°C) preservation medium that enables improved and extended preservation of cells, tissues, and organs
- HypoThermosol™ FRS is uniquely formulated to address the molecular-biological response of cells during the hypothermic preservation process
- HypoThermosol™ FRS includes key ions at concentrations that balance the intracellular state at hypothermic temperatures
- Additional components include pH buffers, energy substrates, free radical scavengers, and osmotic/oncotic stabilizers
Specifications
Hematopoietic Stem and Progenitor Cells; Mesenchymal Stem and Progenitor Cells; Other; Pluripotent Stem Cells | |
Cryopreservation | |
16 x 10 mL |