missing translation for 'onlineSavingsMsg'
Learn More

Hypophosphorous acid solution, 50 wt. % in H2O, Honeywell Fluka™

Product Code. 15641970 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
4 kg
Packaging:
Poly Bottle
Unit Size:
4kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15641970 4 kg Poly Bottle 4kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15641970 Supplier Honeywell-Fluka Supplier No. 2149064KG

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 6303-21-5, 7732-18-5
Boiling Point 108°C
CAS Min % ≥50.0000%
CAS Max % <70.0000%
Density 1.206 g/mL at 25 °C
Packaging Poly Bottle
Quantity 4 kg
Molecular Formula H3O2P
Linear Formula C6H14
MDL Number MFCD02183592
UN Number UN3264
Synonym hypophosphite ion, hypophosphorus acid, phosphinic acid, ion 1-, hydroxyphosphanone, oxo-, e5-phosphanol, hydroxidooxidophosphorus, perfluoroalkyl c6-c12 phosphonic acid, diperfluoroalkyl c6-c12 phosphinic acid, hydroxy-, e5-phosphanone, perfluoro-c6-c12 ethyl hydrogen phosphate
InChI Key ACVYVLVWPXVTIT-UHFFFAOYSA-N
SMILES O[PH2]=O
IUPAC Name phosphinic acid
Molecular Weight (g/mol) 66.00
PubChem CID 22497
ChEBI CHEBI:36363
Formula Weight 66.0
Chemical Name or Material Hypophosphorous acid solution, 50 wt.% in H2O
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.