Learn More
Nitritex™ Hypo-Allergenic, Latex-Free Gloves
Description
The hypoallergenic accelerator-free and latex-free Profile PNHS™ is the ideal choice for those with sensitive skin. The sterile, extra length, nitrile surgical glove is anatomically shaped to enhance comfort and avoid wearer fatigue. The PNHS offers excellent chemical protection and has good puncture resistance. Measuring 300mm/12” the PNHS provides extra coverage than standard surgical gloves and features a beaded cuff to ensure stability on the arm.
Specifications
Specifications
| Color | White |
| Material | Nitrile |
| Product Type | Gloves |
| Size | 6 |
| Sterility | Sterile |
| Quantity | 50Pack |
| Length (Metric) | 300 mm |