Learn More
Thermo Scientific™ Hypersil™ MOS-2 (C8) Columns, 5μm Particle Size
Product Code. 11567471 Shop All Thermo Scientific Products
Click to view available options
Length (Metric):
30 mm
125 mm
150 mm
Diameter (Metric):
2.1 mm
4.6 mm
Unit Size:
Each
Product Code. 11567471
Brand: Thermo Scientific™ 30305034630
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Have a monolayer coverage of C8 alkyl chain chemically bonded onto the silica surface for a reproducible and efficient stationary phase
  • Reliable, less retentive phase
  • Long column lifetimes, even under basic conditions
  • Manufactured under ISO 9001-2000 quality guidelines
  • Affordable
Compliance
Manufactured under ISO 9001-2000 quality guidelines.
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) 4.6 mm
Length (Metric) 30 mm
Particle Size 5 μm
Pore Size 120 Å
USP Type L7
Stationary Phase C8
Packing Material Silica
Carbon Load 6.5%
Column Type Reversed Phase
Phase Reversed Phase